Learn More
Apexbio Technology LLC GSK126, 100mg. Cas: 1346574-57-9 MFCD: MFCD23381067. EZH2 inhibitor
SDP
Supplier:  Apexbio Technology LLC A3446100
Encompass_Preferred
GSK126 is an inhibitor of EZH2 with Ki Value of 93 pM [1].Over-expression of EZH2 has been reported to be correlated with cancer progression and poor prognosis, high grade and high stage in several tumor types. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4802