Learn More
Medchemexpress LLC HY-13470 10mg Medchemexpress, GSK126 CAS:1346574-57-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1347010MG
Encompass_Preferred
Medchemexpress, HY-13470 10mg GSK126 CAS:1346574-57-9 GSK126 (GSK2816126A) is a potent, highly selective inhibitor of EZH2 methyltransferase with an IC50 of 9.9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8999