Learn More
Medchemexpress LLC HY-13470 200mg , GSK126 EZH2 inhibitor;GSK2816126A CAS:1346574-57-9 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13470200MG
Encompass_Preferred
Medchemexpress, HY-13470 200mg GSK126 EZH2 inhibitor;GSK2816126A CAS:1346574-57-9 Formula:C31H38N6O2 IC50: 9.9 nM (EZH2) Purity:98% GSK126 is a potent, highly selective inhibitor of EZH2 methyltransferase activity with IC 50 of 9.9 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2519