Learn More
Selleck Chemical LLC GSK1292263 10mg 1032823-75-8
SDP
Supplier:  Selleck Chemical LLC S214910MG
Encompass_Preferred
GSK1292263 is a novel GPR119 agonist, showing potential for the treatment of type 2 diabetes. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-848-8