Learn More
Medchemexpress LLC Gsk1324726a | 1300031-52-0 | 99.9% | 434.91 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13960100MG
Encompass_Preferred
GSK1324726A, also known as I-BET726, is a novel, potent, and selective inhibitor of BET proteins. It demonstrates high affinity for BRD2, BRD3, and BRD4, and has shown significant inhibition of neuroblastoma tumor growth both in vitro and in vivo.
  • Potent and selective inhibitor of BET proteins
  • High affinity for BRD2 (IC50=41 nM), BRD3 (IC50=31 nM), and BRD4 (IC50=22 nM)
  • Causes potent growth inhibition and cytotoxicity in neuroblastoma cell lines
  • Inhibits neuroblastoma tumor growth in models
Catalog No. 50-003-48929