Learn More
Medchemexpress LLC GSK1702934A | 924377-85-5 | 99.1% | 395.52 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY111098100MG
Encompass_Preferred
GSK1702934A is a selective TRPC3 agonist that modulates cardiac contractility and arrhythmogenesis by activating TRPC3. In vitro, it induces TRPC3/6-currents in HEK293 cells and promotes NCX currents in TRPC3-overexpressing myocytes. In vivo, it transiently increases blood pressure in Sprague Dawley rats.
  • Modulates cardiac contractility and arrhythmogenesis
  • Induces TRPC3/6-currents in HEK293 cells
  • Promotes NCX currents in TRPC3-overexpressing myocytes
  • Increases blood pressure in rats
  • Suitable for research use only
Catalog No. 50-003-44102