Learn More
Medchemexpress LLC GSK180736A | 817194-38-0 | 98.8% | 365.36 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1899050MG
Encompass_Preferred
GSK180736A is a potent Rho-associated coiled-coil kinase 1 (ROCK1) inhibitor with an IC50 of 100 nM. It is also a selective and ATP-competitive G protein-coupled receptor kinase 2 (GRK2) inhibitor with an IC50 of 0.77 μM. Structurally similar to paroxetine, it shows more potency and selectivity for GRK2 than other GRKs. ROCK1 is a potential therapeutic target in cardiovascular diseases like hypertension, and GSK180736A is a weak inhibitor of PKA with an IC50 of 30 μM.
  • Potent rho-associated coiled-coil kinase 1 (ROCK1) inhibitor (IC50 = 100 nM)
  • Selective and ATP-competitive G protein-coupled receptor kinase 2 (GRK2) inhibitor (IC50 = 0.77 μM)
  • Structurally similar to paroxetine
  • More than 100-fold selectivity over other GRKs
  • Potential therapeutic agent for cardiovascular diseases such as hypertension
  • Weak PKA inhibitor (IC50 = 30 μM)
Catalog No. 50-003-86130