Learn More
Selleck Chemical LLC GSK189254A E7924-5MG
SDP
Supplier:  Selleck Chemical LLC E79245MG
Encompass_Preferred
GSK189254A (GSK189254 free base) is a novel potent and selective histamine H3 receptor antagonist with pKi values of 9 59-9 90 and 8 51-9 17 for human and rat H3 respectively
Catalog No. 50-000-42168