Learn More
Medchemexpress LLC HY-10524 10mg , GSK1904529A CAS:1089283-49-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1052410MG
Encompass_Preferred
HY-10524 10mg GSK1904529A CAS: 1089283-49-7 GSK1904529A is a potent, selective, orally active, and ATP-competitive inhibitor of insulin-like growth factor-1 receptor (IGF-1R) and insulin receptor (IR), with IC50s of 27 and 25 nM, respectively. GSK1904529A shows poor activity (IC50>1 μM) in 45 other serine/threonine and tyrosine kinases. GSK1904529A exhibits anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1263