Learn More
Medchemexpress LLC HY-120691A 5mg Medchemexpress, GSK205 CAS:1263068-83-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120691A5MG
Encompass_Preferred
Medchemexpress, HY-120691A 5mg GSK205 CAS:1263068-83-2 GSK205 is a potent, selective TRPV4 antagonist with an IC50 of 4.19  μM for inhibiting TRPV4-mediated Ca2+ influx. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9611