Learn More
Medchemexpress LLC HY-100498 2mg Medchemexpress, GSK2256098 CAS:1224887-10-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1004982MG
Encompass_Preferred
Medchemexpress, HY-100498 2mg GSK2256098 CAS:1224887-10-8 GSK2256098 is a selective FAK kinase inhibitor, which inhibits growth and survival of pancreatic ductal adenocarcinoma cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1855