Learn More
Medchemexpress LLC GSK2292767 | 1254036-66-2 | C24H28N6O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY152805MG
Encompass_Preferred
GSK2292767 is a potent and selective inhibitor of PI3Kδ, exhibiting a pIC50 of 10.1 and over 500-fold selectivity compared to other PI3K isoforms. It is suitable for respiratory disease research. In vivo studies have shown high clearance (50 mL/min/kg) and low oral bioavailability (F < 2%) in rats. It does not impact QT interval, Tpe, or QRS, and carries no significant risk of TdP arrhythmias in rabbit cardiac ventricular wedge assays.
  • Potent and selective PI3Kδ inhibitor
  • Greater than 500-fold selectivity over other PI3K isoforms
  • Useful for respiratory disease research
  • High clearance in rat PK studies
  • Low oral bioavailability in rat PK studies
  • No significant risk of TdP arrhythmias
  • Protects against eosinophil recruitment in a rat model of inflammation
Catalog No. 50-003-72968