Learn More
Medchemexpress LLC GSK256066 Trifluoroacetate | 1415560-64-3 | 99.8% | C29H27F3N4O7S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7006910MM/1ML
Encompass_Preferred
GSK256066 Trifluoroacetate is a selective and high-affinity phosphodiesterase 4 (PDE4) inhibitor, with an IC50 of 3.2 pM for PDE4B. It is developed for the research of chronic obstructive pulmonary disease.
  • Selective and high-affinity phosphodiesterase 4 (PDE4) inhibitor
  • IC50 of 3.2 pM for PDE4B
  • Designed for inhaled administration
  • Highly selective for PDE4
  • Inhibits PDE4 isoforms A-D with equal affinity
  • Inhibits tumor necrosis factor alpha production by lipopolysaccharide (LPS)-stimulated human peripheral blood monocytes
  • Inhibits eosinophil number in bronchoalveolar lavage in animal models
  • Inhibits LPS-induced pulmonary neutrophilia
Catalog No. 50-004-34365