Learn More
Medchemexpress LLC GSK256066 Trifluoroacetate | 1415560-64-3 | 99.8% | C29H27F3N4O7S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY7006950MG
Encompass_Preferred
GSK256066 Trifluoroacetate is a selective and high-affinity phosphodiesterase 4 (PDE) inhibitor, with an IC50 of 3.2 pM for PDE4B. It is developed for the research of chronic obstructive pulmonary disease.
  • Selective and high-affinity phosphodiesterase 4 (PDE) inhibitor
  • IC50 of 3.2 pM for PDE4B
  • Developed for the research of chronic obstructive pulmonary disease
  • Exceptionally high-affinity inhibitor of PDE4 designed for inhaled administration
  • Highly selective for PDE4
  • Inhibits PDE4 isoforms A-D with equal affinity
  • Inhibits tumor necrosis factor α production by LPS-stimulated human peripheral blood monocytes with IC50 of 0.01 nM
  • Inhibits eosinophil number in BAL in LPS- and OVA-induced acute pulmonary inflammation rat models
  • Inhibits LPS-induced pulmonary neutrophilia
Catalog No. 50-004-34227