Learn More
TARGETMOL CHEMICALS INC GSK256073 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T154325MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. GSK256073 is an orally active GPR109A agonist. GSK256073 also is a long-lasting and non-flushing HCA2 (hydroxy-carboxylic acid receptor 2) full agonist (pEC50 7.5). GSK256073 acutely improves glucose homeostasis via inhibition of lipolysis. Purity 98.03%
Catalog No. 50-287-8251