Learn More
Selleck Chemical LLC GSK2606414
SDP
Supplier:  Selleck Chemical LLC S730710MM/1ML
Encompass_Preferred
GSK2606414 is an orally available potent and selective PERK inhibitor with IC50 of 0 4 nM displaying at least 100-fold selectivity over the other EIF2AKs assayed GSK2606414 impairs GANT-61 induced autophagy in NB cells with MYCN amplification GSK2606414 exacerbates ER stress-induced apoptosis in HCT116 cells while reduces the apoptosis in SIL1 KD HeLa cells
Catalog No. 50-000-00043