Learn More
Medchemexpress LLC GSK2606414 | 1337531-36-8 | 99.9% | 451.44 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY18072
Encompass_Preferred
GSK2606414 is a cell-permeable and orally available protein kinase R-like endoplasmic reticulum (ER) kinase (PERK) inhibitor with an IC50 of 0.4 nM. It inhibits PERK activation in cells and has been shown to inhibit the growth of a human tumor xenograft in mice.
  • Cell-permeable
  • Orally available
  • Potent and selective first-in-class inhibitor of protein kinase R (PKR)-like endoplasmic reticulum kinase (PERK)
  • Inhibits PERK activation in cells
  • Inhibits the growth of a human tumor xenograft in mice
Catalog No. 50-003-85456