Learn More
Medchemexpress LLC GSK2636771 | 1372540-25-4 | C22H22F3N3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1524550MG
Encompass_Preferred
GSK2636771 is a potent, selective, and orally bioavailable inhibitor of PI3Kβ with a Ki of 0.89 nM and an IC50 of 5.2 nM. It demonstrates high selectivity, showing 900-fold selectivity over p110α and p110γ, and 10-fold selectivity over p110δ isoforms. In cellular assays, GSK2636771 treatment significantly decreases cell viability in p110β-reliant PTEN-deficient PC3 prostate and BT549 and HCC70 breast cancer cell lines. Inhibition of p110β leads to a marked decrease of AKT phosphorylation in these cancer cell lines.
  • Potent, selective, and orally bioavailable PI3Kβ inhibitor
  • High selectivity over p110α, p110γ, and p110δ isoforms
  • Decreases cell viability in p110β-reliant cancer cell lines
  • Leads to a marked decrease of AKT phosphorylation in cancer cell lines
  • Useful for research in cancer and related signaling pathways
Catalog No. 50-003-72973