Learn More
Selleck Chemical LLC GSK269962A HCl 5mg 2095432-71-4 GSK269962B, GSK269962, GSK 269962
SDP
Supplier:  Selleck Chemical LLC S76875MG
Encompass_Preferred
GSK269962A HCl (GSK269962B, GSK269962) is a selective ROCK(Rho-associated protein kinase) inhibitor with IC50 values of 1.6 and 4 nM for ROCK1 and ROCK2, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5134