Learn More
TARGETMOL CHEMICALS INC GSK270822A 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T274675MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK270822A is a selective ROCK1 inhibitor. GSK270822A exhibits IC50 of 9nM 1100nM 1550nM for ROCK1 RSK1 p70S6K respectively. purity: 98%
Catalog No. 50-258-0610