Learn More
Selleck Chemical LLC GSK2795039 S8974-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S897410MM/1ML
Encompass_Preferred
GSK2795039 is an inhibitor of NADPH oxidase 2 (NOX2) with pIC50 of 6 57 for NOX2-mediated activation of HRP/Amplex Red GSK2795039 inhibits reactive oxygen species (ROS) production NADPH consumption and reduces apoptosis
Catalog No. 50-000-25235