Learn More
Medchemexpress LLC GSK2837808A | 1445879-21-9 | MFCD28133406 | 10 mM * 1 mL in DMSO
SDP
Supplier:  Medchemexpress LLC HY10068110 MM 1 ML
Encompass_Preferred
GSK2837808A | Putity: >99.0% | MW: 649.62 | 1445879-21-9 | MFCD28133406 | 10 mM * 1 mL in DMSO
Catalog No. NC1711267