Learn More
Selleck Chemical LLC GSK2879552 2HCl 5mg 1902123-72-1
SDP
Supplier:  Selleck Chemical LLC S77965MG
Encompass_Preferred
GSK2879552 2HCl is a potent, selective, orally bioavailable, irreversible LSD1 inhibitor with Kiapp of 1.7 ?M. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5261