Learn More
Medchemexpress LLC GSK2983559 free acid | 1579965-12-0 | 99.49% | 538.53 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112038
Encompass_Preferred
GSK2983559 free acid (compound 3) is an orally active and potent receptor interacting protein 2 (RIP2) kinase inhibitor. It can block many proinflammatory cytokine responses in vivo and in human inflammatory bowel disease explant samples. This product is intended for research use only.
  • Blocks MDP-induced IL-8 production in THP-1 cells.
  • Suppresses serum IL-6 levels in vivo.
Catalog No. 50-004-39711