Learn More
Medchemexpress LLC GSK3β inhibitor II | 478482-75-6 | 99.5% | 395.22 g·mol⁻¹ | C14H10IN3OS | 5 MG

Supplier: Medchemexpress LLC HY146795MG
GSK3β inhibitor II is a small-molecule inhibitor of glycogen synthase kinase 3 beta (GSK3β) intended for research use. Supplied as a high-purity solid, it is used in studies of GSK3β-mediated signaling and in models relevant to neurodegenerative disease research.
- High purity suitable for research applications (reported 99.49%).
- Compact 5 mg pack size for small-scale experiments.
- Molecular formula: C14H10IN3OS; molecular weight: 395.22 g·mol⁻¹.
- Provided with supporting documents: data sheet, COA, and SDS.
- Suitable for in vitro signaling and neurobiology studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.