Learn More
Cayman Chemical GSK3 InhIbItr 1 10mg
SDP
Supplier:  Cayman Chemical 4210710 MG
Encompass_Preferred
A GSK3β inhibitor (IC50 = 4.19 nM); decreases blood glucose levels in an OGTT in a rat model of high-fat diet and STZ-induced diabetes at 50 mg/kg
Catalog No. 50-294-3011