Learn More
Cayman Chemical GSK3 InhIbItr II 10mg
SDP
Supplier:  Cayman Chemical 1907710 MG
Encompass_Preferred
A potent inhibitor of GSK3β (IC50 = 390 nM) that does not inhibit GSK-3α
Catalog No. 50-298-4282