Learn More
Sino Biological Peptide Substrates: GSK3 Substrate
Supplier:  Sino Biological G5058
Encompass_Preferred
The GSK3 sub peptide sequence (YRRAAVPPSPSLSRHSSPHQ(pS)EDEEE) is based on human muscle glycogen synthase 1 (amino acid 636-661).
Catalog No. 50-256-9062