Learn More
Medchemexpress LLC HY-111946 10mg Medchemexpress, GSK3145095 CAS:1622849-43-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11194610MG
Encompass_Preferred
Medchemexpress, HY-111946 10mg GSK3145095 CAS:1622849-43-7 GSK3145095 is a RIP1 kinase inhibitor with an IC50 of 6.3 nM . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8990