Learn More
Medchemexpress LLC Gsk3145095 | 1622849-43-7 | 99.4% | 397.38 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111946
Encompass_Preferred
GSK3145095 is a potent RIP1 kinase inhibitor with an IC50 of 6.3 nM. This compound is intended for research applications only and is not suitable for patient use.
  • Functions as a RIP1 kinase inhibitor
  • Exhibits an IC50 of 6.3 nM
  • Suitable for research use
Catalog No. 50-004-38725