Learn More
Selleck Chemical LLC GSK3368715 3HCl S8858-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S885810MM/1ML
Encompass_Preferred
GSK3368715 3HCl is a potent inhibitor of type I protein arginine methyltransferases (PRMT) that inhibits PRMT1 3 4 6 and 8 with Kiapp vaules ranging from 1 5 to 81 nM
Catalog No. 50-000-25415