Learn More
Medchemexpress LLC HY-13500 50mg , GSK343 CAS:1346704-33-3 Purity:>98%
Supplier: Medchemexpress LLC HY1350050MG
HY-13500 50mg GSK343 CAS: 1346704-33-3 GSK343 is a highly potent and selective EZH2 inhibitor with an IC50 of 4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.