Learn More
TARGETMOL CHEMICALS INC GSK369796 Dihydrochloride 2MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T71952MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. GSK369796 Dihydrochloride (N-tert-butylisoquine),is an anti-malaria drug candidate. is an affordable and effective antimalarial and inhibits hERG potassium ion channel repolarization(IC50 of 7.5 uM) Purity 98%
Catalog No. 50-287-7855