Learn More
TARGETMOL CHEMICALS INC GSK429286A 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T263325MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK429286A (RHO-15) is a specific inhibitor of ROCK1/2 (IC50 14/63 nM). purity: 98%
Catalog No. 50-257-7076