Learn More
Selleck Chemical LLC GSK429286A 50mg 864082-47-3 RHO-15
SDP
Supplier:  Selleck Chemical LLC S147450MG
Encompass_Preferred
GSK429286A (RHO-15) is a selective inhibitor of ROCK1 and ROCK2 with IC50 of 14 nM and 63 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-838-4