Learn More
Medchemexpress LLC Gsk484 (free base) | 1652629-23-6 | 99.1% | 473.57 g/mol | C27H31N5O3 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12360625MG
Encompass_Preferred
GSK484 is a selective, reversible inhibitor of peptidylarginine deiminase 4 (PAD4) used in biochemical and cellular research to probe PAD4-mediated processes such as inflammation and NETosis. The compound is provided as the free base with high reported purity and is intended for research applications.
  • Selective, reversible inhibition of PAD4
  • High purity suitable for biochemical assays
  • Suitable for in vitro and cellular studies
  • Characterized by chemical formula C27H31N5O3 and molecular weight 473.6 g/mol
  • Supplied in small research quantities for experimental use
Catalog No. 50-002-37469