Learn More
Medchemexpress LLC GSK484 hydrochloride | 1652591-81-5 | >98.0% | 510.03 | C27H32ClN5O3 | 1 ML

Supplier: Medchemexpress LLC HY10051410MM 1ML
GSK484 hydrochloride is a selective, reversible inhibitor of protein arginine deiminase 4 (PAD4) used in laboratory research to study PAD4-mediated protein citrullination and neutrophil extracellular trap formation. Supplied as a solid or as a ready-to-use 10 mM solution in DMSO, it is intended for biochemical and cellular assays. For research use only.
- Selective, reversible inhibitor of peptidylarginine deiminase 4 (PAD4).
- Potent biochemical activity with reported IC50 ≈ 50 nM in low-calcium conditions, reduced potency with Ca2+ (~250 nM).
- Available as solid and as a 10 mM solution in DMSO for convenience in assays.
- Molecular formula C27H32ClN5O3 and molecular weight 510.03 g/mol.
- CAS number 1652591-81-5 for unambiguous chemical identification.
- Intended for in vitro research applications; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.