Learn More
Medchemexpress LLC GSK547 200mg | 2226735-55-1 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY114492200MG
Encompass_Preferred
GSK547 is a potent, selective small-molecule inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIPK1) used in preclinical research to investigate necroptosis, inflammation, and macrophage-mediated immune tolerance. The compound has molecular formula C20H18F2N6O, molecular weight 396.39 g/mol, and vendor-reported purity of 99.92%. It is supplied in research quantities and as solution formulations for experimental use.
  • Inhibits RIPK1 with high potency and selectivity.
  • Molecular formula C20H18F2N6O; molecular weight 396.39 g/mol.
  • Vendor-reported high purity (~99.9%).
  • Available as a 200 mg solid and as DMSO solutions (10 mM).
  • Suitable for in vitro and preclinical in vivo research applications.
Catalog No. 50-002-29589