Learn More
TARGETMOL CHEMICALS INC GSK583 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T35375MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. GSK583 is a highly effective and specific RIP2 kinase inhibitor (IC50 5 nM). Purity 98.68%
Catalog No. 50-287-8716