Learn More
Medchemexpress LLC HY-137892 5mg Medchemexpress, GSK620 CAS:2088410-46-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1378925MG
Encompass_Preferred
Medchemexpress, HY-137892 5mg GSK620 CAS:2088410-46-0 GSK620 is a potent and orally active pan-BD2 inhibitor with excellent broad selectivity, developability and in vivo oral pharmacokinetics. GSK620 is highly selective for the BET-BD2 family of proteins, with >200-fold selectivity over all other bromodomains. GSK620 shows an anti-inflammatory phenotype in human whole blood. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1514