Learn More
Medchemexpress LLC Gsk737 | 2748687-95-6 | C20H21N5O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15324150MG
Encompass_Preferred
GSK737 is an inhibitor of BD1 and BD2. It demonstrates low clearance and good solubility and permeability in rat models.
  • Functions as a BD1 and BD2 inhibitor.
  • Exhibits low clearance in rat.
  • Possesses good solubility in rat.
  • Demonstrates good permeability in rat.
Catalog No. 50-003-72612