Learn More
TARGETMOL CHEMICALS INC GSK805 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T73885MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. GSK805 is a potent, orally bioavailable and CNS-penetrant RORgammat inhibitor. Purity 98.02%
Catalog No. 50-287-6831