Learn More
Apexbio Technology LLC GSK962040 hydrochloride 923565-22-4 2mg
SDP
Supplier:  Apexbio Technology LLC B11372
Encompass_Preferred
GSK962040 hydrochloride is a chemically synthesized small-molecule agonist targeting the motilin receptor It is designed to activate motilin receptor signaling pathways thereby modulating gastrointestinal motility GSK962040 hydrochloride exerts its biological activity primarily through selective agonism of the motilin receptor In in vitro and in vivo studies GSK962040 hydrochloride demonstrates agonist activity with a pEC50 value of approximately 7 9 Based on these pharmacological properties GSK962040 hydrochloride holds research potential in preclinical studies of gastrointestinal physiology particularly in exploring motilin receptor-related mechanisms and motility disorders
Catalog No. 50-000-90727