Learn More
Vector Laboratories GSL I - Isolectin B4 (GSL I-B4 , BSL I-B4 ) Biotinylated 0.5 mg
SDP
Supplier:  Vector Laboratories B1205.5
Encompass
Griffonia simplicifolia isolectin (GSL I-B4) contains only B subunits and is a useful marker for endothelial cells from nonprimates, such as mouse, rat, rabbit, and goat. It is also a marker for non-peptidergic unmyelinated primary afferent neurons. This "B"-rich lectin preferentially agglutinates blood group B cells and is specific for α-galactose residues. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
Catalog No. NC9918852