Learn More
Vector Laboratories Griffonia (Bandeiraea) Simplicifolia Lectin I (GSL I, BSL I), Unconjugated 5 mg
SDP
Supplier:  Vector Laboratories L11005
Encompass
Griffonia simplicifolia lectin I (GSL I) is a family of glycoproteins with molecular weights of approximately 114 kDa. Two subunits ("A" and "B") have slightly different molecular weights and combine to form tetrameric structures, resulting in five isolectins. The "A"-rich lectin preferentially agglutinates blood group A erythrocytes and appears to be specific for α-N-acetylgalactosamine residues, while the "B"-rich lectin preferentially agglutinates blood group B cells and is specific for α-galactose residues.
Catalog No. NC0833913