Learn More
Selleck Chemical LLC GsMTx4 1mg
SDP
Supplier:  Selleck Chemical LLC P12051MG
Encompass_Preferred
GsMTx4 (Grammostola spatulata mechanotoxin 4, M-theraphotoxin-Gr1a, M-TRTX-Gr1a) is a spider venom peptide that inhibits cationic mechanosensitive channels (MSCs). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3017