Learn More
Medchemexpress LLC Gstm2 protein, human (his, solution) | >95.0% | 250 UG
SDP
Supplier:  Medchemexpress LLC HYP75152A250UG
Encompass_Preferred
Recombinant human glutathione S-transferase Mu 2 (GSTM2) with an N-terminal His tag supplied as a lyophilized preparation for research use. Expressed in Escherichia coli and purified to >95% purity, this 218-amino-acid protein is provided in a 250 μg vial and formulated with stabilizers for storage and handling.
  • Recombinant human glutathione S-transferase Mu 2 with N-terminal His tag.
  • Expressed in E. coli and purified to greater than 95% by reducing SDS-PAGE.
  • Full-length sequence (M1-K218), suitable for biochemical and binding assays.
  • Lyophilized formulation with trehalose and Tween 80 for enhanced stability.
  • Supplied as a 250 μg vial; store at -20°C and aliquot for long-term storage.
  • For research use only; not for therapeutic or diagnostic use.
Catalog No. 50-002-92794