Learn More
Medchemexpress LLC GSTP1 Human His 100ug
SDP
Supplier:  Medchemexpress LLC HYP72215100UG
Encompass_Preferred
Recombinant human glutathione S-transferase P1 (GSTP1) with an N-terminal His tag, expressed in E. coli and supplied as a highly purified protein for biochemical and structural research. It is suitable for enzyme activity assays, antibody validation, and other applications requiring purified human GSTP1.
  • Recombinant human GSTP1 with N-terminal His tag.
  • Expressed in E. coli and purified to >95% by reducing SDS-PAGE.
  • Approximate molecular weight 25-30 kDa.
  • Available in multiple sizes, including 100 μg.
  • Store frozen at -20°C; reconstituted material is stable at 4°C for 1 week.
  • Recommended reconstitution ≥100 μg/mL in ddH2O and addition of carrier protein for long-term storage.
Catalog No. 50-001-83584