Learn More
ABclonal Technology GSTT1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A23070100UL
Encompass_Preferred
The protein encoded by this gene glutathione S-transferase (GST) theta 1 (GSTT1) is a member of a superfamily of proteins that catalyze the conjugation of reduced glutathione to a variety of electrophilic and hydrophobic compounds Human GSTs can be divided into five main classes alpha mu pi theta and zeta The theta class includes GSTT1 GSTT2 and GSTT2B GSTT1 and GSTT2 GSTT2B share 55% amino acid sequence identity and may play a role in human carcinogenesis The GSTT1 gene is haplotype-specific and is absent from 38% of the population Alternative splicing of this gene results in multiple transcript variants
Catalog No. 50-000-04852