Learn More
Cytek Biosciences Inc Gt aChicken IgY Anti Secondary
Supplier: Cytek Biosciences Inc 358081M002
Product Name: FITC Goat Anti-Chicken IgY (H&L)
Product Class: Antibody
Type: Secondary
Product Name: FITC Goat Anti-Chicken IgY (H&L)
Cytek Catalog Number: 35-8081-M002
Marker: IgY (H&L)
Target Species: Chicken
Reported Cross Reactivity: Chicken
Clonality: Polyclonal
Host Species: Goat
Conjugation: FITC
Size: 2 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.